Assay Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267599.3 | NP_001254528.1 | 292 | 54 | ||
| GenBank mRNA | JV546252.1 | - | 132 | 54 | ||
| JV667406.1 | - | 292 | 54 | |||
More Information
Set Membership: |
|

NRGN
Genomic Map