Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194831.2 | NP_001181760.1 | 1185 | 142 | ||
| XM_028838499.1 | XP_028694332.1 | 2106 | 142 | |||
| XM_028838500.1 | XP_028694333.1 | 1983 | 142 | |||
| XM_028838501.1 | XP_028694334.1 | 1988 | 142 | |||
More Information
Set Membership: |
|

NTF4
Genomic Map