Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114120.2 | XP_014969606.1 | 1596 | 102 | ||
| XM_015114122.2 | XP_014969608.1 | 1135 | 102 | |||
| XM_028832627.1 | XP_028688460.1 | 1063 | 102 | |||
| GenBank mRNA | JU319951.1 | - | 735 | 102 | ||
| JU474101.1 | - | 718 | 102 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CLP1
Genomic Map