Assay Details
| Gene Symbol: DEPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146915.2 | XP_015002401.1 | 291 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015146915.2 transcripts.
Set Membership: |
|

DEPP1
Genomic Map