Assay Details
| Gene Symbol: LSM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266965.2 | NP_001253894.1 | 271 | 110 | ||
| XM_015139278.2 | XP_014994764.1 | 455 | 110 | |||
| XM_015139279.2 | XP_014994765.1 | 462 | 110 | |||
| XM_015139280.2 | XP_014994766.1 | 350 | 110 | |||
| GenBank mRNA | JU319787.1 | - | 316 | 110 | ||
| JV046059.1 | - | 273 | 110 | |||
More Information
Set Membership: |
|

LSM6
Genomic Map