Assay Details
| Gene Symbol: SOX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193732.3 | NP_001180661.1 | 924 | 83 | ||
| XM_015121305.2 | XP_014976791.1 | 1062 | 83 | |||
| XM_028837090.1 | XP_028692923.1 | 1062 | 83 | |||
More Information
Set Membership: |
|

SOX21
Genomic Map