Assay Details
Gene Symbol: OR2H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194222.2 | NP_001181151.2 | 69 | 126 | ||
GenBank mRNA | JV489100.1 | - | 277 | 126 | ||
JV527766.1 | - | 310 | 126 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194222.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
