Assay Details
| Gene Symbol: PRND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149366.2 | XP_015004852.1 | 86 | 157 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015149366.2 transcripts.
Set Membership: |
|

PRND
Genomic Map