Assay Details
| Gene Symbol: MRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266892.1 | NP_001253821.1 | 322 | 79 | ||
| GenBank mRNA | JU334225.1 | - | 323 | 79 | ||
| JU475339.1 | - | 328 | 79 | |||
| JV527798.1 | - | 322 | 79 | |||
| JV713284.1 | - | 120 | 79 | |||
More Information
Set Membership: |
|

MRM2
Genomic Map