Assay Details
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114259.4 | XP_001114259.2 | 3091 | 122 | ||
| XM_028844543.1 | XP_028700376.1 | 1313 | 122 | |||
More Information
Set Membership: |
|

KLF15
Genomic Map