Assay Details
| Gene Symbol: NDUFAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260754.1 | NP_001247683.1 | 288 | 67 | ||
| GenBank mRNA | JU332311.1 | - | 321 | 67 | ||
| JU475282.1 | - | 284 | 67 | |||
| JV045453.1 | - | 288 | 67 | |||
| JV567093.1 | - | 317 | 67 | |||
| JV638897.1 | - | 323 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFAF4
Genomic Map