Assay Details
| Gene Symbol: SMUG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194757.1 | NP_001181686.1 | 426 | 130 | ||
| XM_001109204.4 | XP_001109204.1 | 620 | 130 | |||
| XM_015151814.2 | XP_015007300.1 | 1042 | 130 | |||
| XM_015151815.2 | XP_015007301.1 | 613 | 130 | |||
| XM_015151816.2 | XP_015007302.1 | 1035 | 130 | |||
| XM_028828872.1 | XP_028684705.1 | 483 | 130 | |||
| GenBank mRNA | JU322874.1 | - | 300 | 130 | ||
| JU473614.1 | - | 302 | 130 | |||
| JV567040.1 | - | 306 | 130 | |||
More Information
Set Membership: |
|

SMUG1
Genomic Map