Assay Details
| Gene Symbol: SOX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125258.2 | XP_014980744.2 | 591 | 152 | ||
| GenBank mRNA | JU323906.1 | - | 539 | 152 | ||
| JV566339.1 | - | 528 | 152 | |||
| JV600984.1 | - | 544 | 152 | |||
More Information
Set Membership: |
|

SOX8
Genomic Map