Assay Details
| Gene Symbol: MEA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194869.1 | NP_001181798.1 | 508 | 86 | ||
| XM_001087765.4 | XP_001087765.2 | 531 | 86 | |||
| Gene Symbol: PPP2R5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU335298.1 | - | 3224 | 86 | ||
| JU471102.1 | - | 3220 | 86 | |||
| JV045007.1 | - | 3223 | 86 | |||
| JV566389.1 | - | 3232 | 86 | |||
| JV600899.1 | - | 3185 | 86 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MEA1
Genomic Map