Assay Details
| Gene Symbol: PHYHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266549.1 | NP_001253478.1 | 1017 | 151 | ||
| XM_015144900.2 | XP_015000386.1 | 657 | 151 | |||
| XM_028852270.1 | XP_028708103.1 | 1039 | 151 | |||
| XM_028852271.1 | XP_028708104.1 | 657 | 151 | |||
| GenBank mRNA | JU337332.1 | - | 1017 | 151 | ||
| JV564374.1 | - | 1027 | 151 | |||
More Information
Set Membership: |
|

PHYHIP
Genomic Map