Assay Details
| Gene Symbol: ZNF691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137277.2 | XP_014992763.2 | 1499 | 157 | ||
| XM_015137284.2 | XP_014992770.2 | 1351 | 157 | |||
| XM_015137302.2 | XP_014992788.2 | 1044 | 157 | |||
| XM_028835017.1 | XP_028690850.1 | 2299 | 157 | |||
| XM_028835019.1 | XP_028690852.1 | 1606 | 157 | |||
| XM_028835021.1 | XP_028690854.1 | 1506 | 157 | |||
| XM_028835024.1 | XP_028690857.1 | 1639 | 157 | |||
| XM_028835025.1 | XP_028690858.1 | 1238 | 157 | |||
| XM_028835026.1 | XP_028690859.1 | 2151 | 157 | |||
| XM_028835031.1 | XP_028690864.1 | 2038 | 157 | |||
| XM_028835035.1 | XP_028690868.1 | 1068 | 157 | |||
| XM_028835037.1 | XP_028690870.1 | 1173 | 157 | |||
| XM_028835042.1 | XP_028690875.1 | 2012 | 157 | |||
| GenBank mRNA | JU344260.1 | - | 1008 | 157 | ||
| JU487626.1 | - | 986 | 157 | |||
More Information
Set Membership: |
|

ZNF691
Genomic Map