Assay Details
| Gene Symbol: MRPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257904.1 | NP_001244833.1 | 344 | 134 | ||
| GenBank mRNA | JU334761.1 | - | 344 | 134 | ||
| JU473741.1 | - | 385 | 134 | |||
| JV567874.1 | - | 404 | 134 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MRPS16
Genomic Map