Assay Details
| Gene Symbol: ACTR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109946.2 | XP_014965432.1 | 1777 | 84 | ||
| GenBank mRNA | JU332403.1 | - | 1288 | 84 | ||
| JU473864.1 | - | 1390 | 84 | |||
| JV045834.1 | - | 1347 | 84 | |||
More Information
Set Membership: |
|

ACTR3
Genomic Map