Assay Details
| Gene Symbol: YY1AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827998.1 | XP_028683831.1 | 631 | 134 | ||
| XM_028828000.1 | XP_028683833.1 | 538 | 134 | |||
| XM_028828006.1 | XP_028683839.1 | 563 | 134 | |||
| XM_028828013.1 | XP_028683846.1 | 744 | 134 | |||
| XM_028828019.1 | XP_028683852.1 | 637 | 134 | |||
| XM_028828026.1 | XP_028683859.1 | 463 | 134 | |||
| XM_028828029.1 | XP_028683862.1 | 666 | 134 | |||
| XM_028828031.1 | XP_028683864.1 | 591 | 134 | |||
| XM_028828034.1 | XP_028683867.1 | 563 | 134 | |||
| XM_028828039.1 | XP_028683872.1 | 567 | 134 | |||
| XM_028828047.1 | XP_028683880.1 | 609 | 134 | |||
| XM_028828055.1 | XP_028683888.1 | 568 | 134 | |||
| XM_028828057.1 | XP_028683890.1 | 593 | 134 | |||
| XM_028828060.1 | XP_028683893.1 | 503 | 134 | |||
| XM_028828064.1 | XP_028683897.1 | 593 | 134 | |||
| XM_028828069.1 | XP_028683902.1 | 592 | 134 | |||
| GenBank mRNA | JU476133.1 | - | 2144 | 134 | ||
| JV585478.1 | - | 1360 | 134 | |||
| Gene Symbol: LOC719341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU335132.1 | - | 2144 | 134 | ||
| JV047459.1 | - | 2149 | 134 | |||
| JV636239.1 | - | 2141 | 134 | |||
More Information
Set Membership: |
|

YY1AP1
Genomic Map