Assay Details
Gene Symbol: PCGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015147673.2 | XP_015003159.1 | 1024 | 106 | ||
XM_015147674.2 | XP_015003160.1 | 1378 | 106 | |||
XM_015147677.2 | XP_015003163.1 | 884 | 106 | |||
XM_028826651.1 | XP_028682484.1 | 884 | 106 | |||
XM_028826652.1 | XP_028682485.1 | 864 | 106 | |||
XM_028826653.1 | XP_028682486.1 | 1710 | 106 | |||
XM_028826654.1 | XP_028682487.1 | 864 | 106 | |||
XM_028826655.1 | XP_028682488.1 | 884 | 106 | |||
GenBank mRNA | JU320328.1 | - | 790 | 106 | ||
JU320329.1 | - | 790 | 106 | |||
JU320330.1 | - | 790 | 106 | |||
JU320331.1 | - | 790 | 106 | |||
JU473191.1 | - | 847 | 106 | |||
JV602535.1 | - | 854 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
