Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002808008.3 | XP_002808054.1 | 1789 | 115 | ||
| XM_015110464.2 | XP_014965950.1 | 1055 | 115 | |||
| XM_015110467.2 | XP_014965953.1 | 1041 | 115 | |||
| XM_028830892.1 | XP_028686725.1 | 1718 | 115 | |||
| XM_028830894.1 | XP_028686727.1 | 1081 | 115 | |||
| XM_028830895.1 | XP_028686728.1 | 1238 | 115 | |||
| XM_028830896.1 | XP_028686729.1 | 1091 | 115 | |||
| XM_028830897.1 | XP_028686730.1 | 1478 | 115 | |||
| XM_028830898.1 | XP_028686731.1 | 974 | 115 | |||
| XM_028830899.1 | XP_028686732.1 | 1414 | 115 | |||
| XM_028830900.1 | XP_028686733.1 | 1203 | 115 | |||
| XM_028830901.1 | XP_028686734.1 | 1536 | 115 | |||
| XM_028830902.1 | XP_028686735.1 | 1009 | 115 | |||
| GenBank mRNA | JU333107.1 | - | 1363 | 115 | ||
| JV703725.1 | - | 1585 | 115 | |||
| JV703726.1 | - | 1585 | 115 | |||
More Information
Set Membership: |
|

ZNF385B
Genomic Map