Assay Details
| Gene Symbol: ZBTB7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093405.4 | XP_001093405.2 | 1864 | 118 | ||
| XM_015121898.2 | XP_014977384.2 | 1704 | 118 | |||
| XM_028838134.1 | XP_028693967.1 | 1689 | 118 | |||
| GenBank mRNA | JV054231.1 | - | 1207 | 118 | ||
| JV636017.1 | - | 1221 | 118 | |||
More Information
Set Membership: |
|

ZBTB7C
Genomic Map