Assay Details
| Gene Symbol: MAPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015142257.2 | XP_014997743.1 | 1995 | 149 | ||
| XM_015142258.2 | XP_014997744.1 | 2028 | 149 | |||
| XM_015142259.2 | XP_014997745.1 | 2018 | 149 | |||
| GenBank mRNA | JU322195.1 | - | 1978 | 149 | ||
| JU471407.1 | - | 1975 | 149 | |||
| JU471408.1 | - | 1975 | 149 | |||
| JV044412.1 | - | 1962 | 149 | |||
| JV566004.1 | - | 1980 | 149 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAPK6
Genomic Map