Assay Details
| Gene Symbol: CHST6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841081.1 | XP_028696914.1 | 575 | 122 | ||
| GenBank mRNA | JU324006.1 | - | 100 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028841081.1 transcripts.
Set Membership: |
|

CHST6
Genomic Map