Assay Details
| Gene Symbol: PHETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152801.2 | XP_015008287.1 | 345 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015152801.2 transcripts.
Set Membership: |
|

PHETA1
Genomic Map