Assay Details
| Gene Symbol: HADHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111868.2 | XP_014967354.1 | 463 | 147 | ||
| XM_015111869.2 | XP_014967355.1 | 409 | 147 | |||
| XM_028831894.1 | XP_028687727.1 | 433 | 147 | |||
| GenBank mRNA | JU485305.1 | - | 305 | 147 | ||
| JV059320.1 | - | 316 | 147 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HADHB
Genomic Map