Assay Details
| Gene Symbol: TMEM98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119048.2 | XP_014974534.1 | 574 | 100 | ||
| XM_015119049.2 | XP_014974535.1 | 525 | 100 | |||
| GenBank mRNA | JU320225.1 | - | 212 | 100 | ||
| JU320226.1 | - | 394 | 100 | |||
| JU473651.1 | - | 288 | 100 | |||
| JV043376.1 | - | 389 | 100 | |||
| JV711613.1 | - | 395 | 100 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM98
Genomic Map