Assay Details
| Gene Symbol: TUBG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261011.1 | NP_001247940.1 | 592 | 151 | ||
| GenBank mRNA | JU471734.1 | - | 394 | 151 | ||
| JV044189.1 | - | 592 | 151 | |||
| JV635408.1 | - | 396 | 151 | |||
| Gene Symbol: LOC100429346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU320333.1 | - | 578 | 151 | ||
| JU471733.1 | - | 394 | 151 | |||
| JV046772.1 | - | 592 | 151 | |||
More Information
Set Membership: |
|

TUBG1
Genomic Map