Assay Details
| Gene Symbol: LOC710412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028848544.1 | XP_028704377.1 | 1264 | 148 | ||
| XM_028848545.1 | XP_028704378.1 | 1320 | 148 | |||
More Information
Set Membership: |
|

LOC710412
Genomic Map