Assay Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258044.1 | NP_001244973.1 | 131 | 151 | ||
| GenBank mRNA | JU472214.1 | - | 131 | 151 | ||
| JV566183.1 | - | 138 | 151 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001258044.1 transcripts.
Set Membership: |
|

HSBP1
Genomic Map