Assay Details
| Gene Symbol: AKR1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001118631.4 | XP_001118631.3 | 150 | 122 | ||
| XM_028825937.1 | XP_028681770.1 | 131 | 122 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AKR1C4
Genomic Map