Assay Details
| Gene Symbol: SLC31A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257506.1 | NP_001244435.1 | 377 | 131 | ||
| GenBank mRNA | JU332952.1 | - | 377 | 131 | ||
| JU332953.1 | - | 377 | 131 | |||
| JU473239.1 | - | 354 | 131 | |||
| JV046212.1 | - | 389 | 131 | |||
| JV642423.1 | - | 378 | 131 | |||
More Information
Set Membership: |
|

SLC31A1
Genomic Map