Assay Details
| Gene Symbol: EIF4A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195661.1 | NP_001182590.1 | 226 | 136 | ||
| XM_015132415.2 | XP_014987901.1 | 2109 | 136 | |||
| XM_015132416.2 | XP_014987902.1 | 292 | 136 | |||
| XM_028843269.1 | XP_028699102.1 | 1794 | 136 | |||
| GenBank mRNA | EG703357.1 | - | 231 | 136 | ||
| JU327120.1 | - | 182 | 136 | |||
| JV535894.1 | - | 189 | 136 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EIF4A2
Genomic Map