Assay Details
| Gene Symbol: DUSP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266548.1 | NP_001253477.1 | 395 | 143 | ||
| GenBank mRNA | JU333262.1 | - | 422 | 143 | ||
| JV046357.1 | - | 395 | 143 | |||
| JV708682.1 | - | 286 | 143 | |||
More Information
Set Membership: |
|

DUSP3
Genomic Map