Assay Details
| Gene Symbol: HSPA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140986.2 | XP_014996472.2 | 1598 | 80 | ||
| GenBank mRNA | JU334300.1 | - | 968 | 80 | ||
| JU475408.1 | - | 1040 | 80 | |||
| JV043757.1 | - | 1143 | 80 | |||
| JV504944.1 | - | 851 | 80 | |||
More Information
Set Membership: |
|

HSPA9
Genomic Map