Assay Details
| Gene Symbol: LOC713572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851424.1 | XP_028707257.1 | 2299 | 133 | ||
| XM_028851426.1 | XP_028707259.1 | 405 | 133 | |||
| GenBank mRNA | JV564947.1 | - | 1200 | 133 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC713572
Genomic Map