Assay Details
| Gene Symbol: METTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116496.4 | XP_001116496.2 | 281 | 94 | ||
| XM_015152140.2 | XP_015007626.1 | 281 | 94 | |||
| GenBank mRNA | JU321158.1 | - | 118 | 94 | ||
| JV045429.1 | - | 126 | 94 | |||
| JV651758.1 | - | 78 | 94 | |||
More Information
Set Membership: |
|

METTL1
Genomic Map