Assay Details
| Gene Symbol: SAE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124334.2 | XP_014979820.1 | 754 | 121 | ||
| XM_015124336.2 | XP_014979822.1 | 738 | 121 | |||
| XM_028839345.1 | XP_028695178.1 | 600 | 121 | |||
| XM_028839346.1 | XP_028695179.1 | 853 | 121 | |||
| XM_028839347.1 | XP_028695180.1 | 595 | 121 | |||
| GenBank mRNA | JU332387.1 | - | 620 | 121 | ||
| JV043206.1 | - | 658 | 121 | |||
| JV568323.1 | - | 725 | 121 | |||
| JV639640.1 | - | 731 | 121 | |||
More Information
Set Membership: |
|

SAE1
Genomic Map