Assay Details
| Gene Symbol: EHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261195.1 | NP_001248124.1 | 1013 | 123 | ||
| XM_015113763.2 | XP_014969249.1 | 1041 | 123 | |||
| XM_015113764.2 | XP_014969250.1 | 1042 | 123 | |||
| GenBank mRNA | JU332645.1 | - | 1063 | 123 | ||
| JU474090.1 | - | 1095 | 123 | |||
| JV043257.1 | - | 1013 | 123 | |||
| JV604173.1 | - | 1147 | 123 | |||
More Information
Set Membership: |
|

EHD1
Genomic Map