Assay Details
| Gene Symbol: CDC37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105596.4 | XP_001105596.2 | 502 | 93 | ||
| GenBank mRNA | JU320783.1 | - | 190 | 93 | ||
| JU470466.1 | - | 194 | 93 | |||
| JV529694.1 | - | 191 | 93 | |||
| JV566270.1 | - | 184 | 93 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDC37
Genomic Map