Assay Details
| Gene Symbol: CLIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261607.1 | NP_001248536.1 | 338 | 150 | ||
| XM_015131177.2 | XP_014986663.1 | 387 | 150 | |||
| GenBank mRNA | JU319666.1 | - | 360 | 150 | ||
| JU473589.1 | - | 325 | 150 | |||
| JV044555.1 | - | 338 | 150 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015131177.2 transcripts.
Set Membership: |
|

CLIC4
Genomic Map