Assay Details
| Gene Symbol: SERBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141988.2 | XP_014997474.1 | 1282 | 97 | ||
| XM_015141998.2 | XP_014997484.1 | 1264 | 97 | |||
| XM_015142008.2 | XP_014997494.1 | 1237 | 97 | |||
| XM_015142018.2 | XP_014997504.1 | 1219 | 97 | |||
| GenBank mRNA | JU327934.1 | - | 196 | 97 | ||
| JU327935.1 | - | 151 | 97 | |||
| JU484249.1 | - | 243 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SERBP1
Genomic Map