Assay Details
| Gene Symbol: FAM8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258124.2 | NP_001245053.1 | 1068 | 159 | ||
| GenBank mRNA | JU475900.1 | - | 1068 | 159 | ||
| JV047302.1 | - | 997 | 159 | |||
| JV638121.1 | - | 1035 | 159 | |||
| JV716582.1 | - | 1008 | 159 | |||
More Information
Set Membership: |
|

FAM8A1
Genomic Map