Assay Details
| Gene Symbol: LPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144880.2 | XP_015000366.1 | 1091 | 74 | ||
| GenBank mRNA | JU334492.1 | - | 630 | 74 | ||
| JU475586.1 | - | 613 | 74 | |||
More Information
Set Membership: |
|

LPL
Genomic Map