Assay Details
| Gene Symbol: MRPS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135485.2 | XP_014990971.1 | 1173 | 113 | ||
| GenBank mRNA | JU476851.1 | - | 723 | 113 | ||
| JV721404.1 | - | 747 | 113 | |||
More Information
Set Membership: |
|

MRPS15
Genomic Map