Assay Details
| Gene Symbol: PBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136041.2 | XP_014991527.2 | 1316 | 71 | ||
| GenBank mRNA | JU328119.1 | - | 945 | 71 | ||
| JU475792.1 | - | 1207 | 71 | |||
| JV043523.1 | - | 1079 | 71 | |||
| JV618201.1 | - | 1280 | 71 | |||
More Information
Set Membership: |
|

PBX2
Genomic Map