Assay Details
| Gene Symbol: RFPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150039.2 | XP_015005525.2 | 347 | 111 | ||
| Gene Symbol: LOC718691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150112.2 | XP_015005598.2 | 246 | 111 | ||
| XM_028828757.1 | XP_028684590.1 | 642 | 111 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015150112.2 transcripts.
Set Membership: |
|

RFPL1
Genomic Map