Assay Details
Gene Symbol: GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193715.2 | NP_001180644.1 | 561 | 76 | ||
XM_015127850.2 | XP_014983336.1 | 2099 | 76 | |||
XM_015127852.2 | XP_014983338.1 | 2060 | 76 | |||
GenBank mRNA | JU332811.1 | - | 922 | 76 | ||
JU332813.1 | - | 1047 | 76 | |||
JU474220.1 | - | 1151 | 76 | |||
JU474221.1 | - | 922 | 76 | |||
JU474223.1 | - | 1151 | 76 | |||
JV046114.1 | - | 1047 | 76 | |||
JV046115.1 | - | 1047 | 76 | |||
JV672232.1 | - | 1116 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
