Assay Details
| Gene Symbol: LOC699486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126193.2 | XP_014981679.2 | 151 | 77 | ||
| XM_028841465.1 | XP_028697298.1 | 252 | 77 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015126193.2, XM_028841465.1 transcripts.
Set Membership: |
|

LOC699486
Genomic Map