Assay Details
| Gene Symbol: LOC719765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835366.1 | XP_028691199.1 | 2930 | 116 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC719765| Gene Symbol: LOC719765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835366.1 | XP_028691199.1 | 2930 | 116 | ||
Set Membership: |
|